Skip to Content
Merck
All Photos(1)

Key Documents

23771

Supelco

KIT-CAPILLARY GAS FLOW BUBBLE METER, STAND INCLUDED

Synonym(s):

Manual Bubble Flowmeter (Capillary Version)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$309.00

$309.00


Estimated to ship on11 April 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$309.00

About This Item

UNSPSC Code:
41112500

$309.00


Estimated to ship on11 April 2025


Request a Bulk Order

General description

Bubble flowmeter is a simple and precise way to measure flow rates of gases.[1] It determines the volumetric flow rate by measuring the time taken by the gas stream to move a soap bubble through a specific volume.
This 0.5 mL bubble flowmeter allows the true split ratio to be determined by measuring actual flow rate. The flow tube is graduated in 0.01 mL increments to allow for the low flows typical of capillary GC applications. The flow tube is held by two magnetic clamps so the unit can be attached to any metal surface for easy storage. A version that includes a sampling stand is offered for added portability.

Application

Bubble flowmeter may be used to manually control the flow at the system outlet during analysis of perfluorobutane (PFB) in the exhaled rat air using respiration-metabolism chamber system and a GC-MS method.[2] Bubble flowmeters can be used to check splitter or detector flow or to calibrate personal pumps for accurate air sampling.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The accuracy of the bubble meter method for gas flow measurements.
Levy A.
Journal of Scientific Instruments, 41 (7), 449-453 (1964)
Steinar Uran et al.
Journal of pharmaceutical and biomedical analysis, 39(3-4), 746-751 (2005-06-21)
Sonazoid is a new contrast agent for ultrasound imaging comprising an aqueous suspension of lipid-stabilised perfluorobutane (PFB) gas microbubbles. A respiration-metabolism chamber system was developed to collect exhaled air following intravenous administration of Sonazoid to rats. Analysis of PFB in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service