Skip to Content
Merck
All Photos(1)

Documents

22891

Chart Paper for Recorder

Varian 03-917650-01 (OEM), chart type, black thermal, pkg of 12 ea

Synonym(s):

THERMAL CHART PAPER Varian® 3000 SERIES 12 ROLLS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
14111500

feature

chart type black thermal

packaging

pkg of 12 ea

manufacturer/tradename

Varian 03-917650-01 (OEM)

compatibility

for use with Varian 3000

Looking for similar products? Visit Product Comparison Guide

General description

Main purpose of a recorder is to record electrical and non-electrical quantity as a function of time. Graphic recorders use elementary rudiments like chart paper for displaying and storing the physical quantity and pen for marking the variation in physical quantity. Chart paper is passed through the recorder at constant rate. The recorder itself becomes a device for measuring retention time. A certain number of inches or centimetres of chart paper corresponds to a certain number of minutes.

Application

Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.

Legal Information

Varian is a registered trademark of Varian Medical Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service