Skip to Content
Merck
All Photos(1)

Key Documents

22350-U

Chart Paper for Recorder

chart type, blank roll

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$157.00

$157.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$157.00

About This Item

UNSPSC Code:
12352200
NACRES:
SB.54

$157.00


Check Cart for Availability

Request a Bulk Order

feature

chart type blank roll

packaging

pkg of 1 ea

manufacturer/tradename

Agilent/HP 5181-1219 (OEM)

compatibility

for use with Agilent/HP 3394, 3395, or 3396

Looking for similar products? Visit Product Comparison Guide

Application

Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.[1]

Other Notes

Equivalent to Manufacturer′s Chart No. 5181-1219.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service