Key Documents
22063
Compact Adapter
About This Item
UNSPSC Code:
41115717
description
compact reducer
packaging
pkg of 1 ea
General description
Use as a 1/4 in. to 1/16 in. Swagelok reducer,
or as a 1/4 in. to 1/16 in. capillary I.D. reducing ferrule.
Also fits Varian capillary injection ports. Stainless steel body. Includes 1/4 in. M-2A ferrule. Purchase appropriate capillary ferrules separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)