Skip to Content
Merck
All Photos(1)

Key Documents

21688-U

Supelco

Glass Wool

Pesticide Grade (Silanized), pkg of 100 g

Synonym(s):

Silanized Glass Wool

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$1,130.00

$1,130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$1,130.00

About This Item

EC Number:
UNSPSC Code:
41121800
NACRES:
SB.54

$1,130.00


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 100 g

Looking for similar products? Visit Product Comparison Guide

General description

Glass wool is made up of short glass fibres and is non-flammable in nature. It is widely considered as a thermal insulation material in ships, automobiles, air conditioning ducts and water supply pipes. Furthermore, it is also used as noise absorbing material in concert halls, studios and movie theatres. Above all that it is non carcinogenic to humans.[1]
Wool plugs are often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column. They can also be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain adsorbent beds.

Application

Silanized glass wool gas chromatographic packed column maybe used in an experimental procedure for measuring and collecting volatiles from edible oils.[2]

Other Notes

Pesticide Grade (Silanized) for applications that involve active analytes, such as organochlorine and/or organophosphorous pesticides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imanaka Y.
Advanced Ceramic Technologies & Products (2012)
Volatiles and oil quality
Jackson HW and Giacherio DJ.
Journal of the American Oil Chemists' Society, 54, 458-460 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service