Key Documents
21031
Supelco
Filter Cartridge Assembly
Select a Size
1 EA
$142.00
$142.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$142.00
About This Item
UNSPSC Code:
12190000
$142.00
Check Cart for Availability
Request a Bulk Order
Agency
EPA IP-8,TO-10A
packaging
pkg of 1 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Filter Cartridge Assembly includes 1 O-ring (a), 1 filter cartridge body (b), 1 stainless steel screen (c), 1 quartz filter (d), 1 filter ring (e), 1 cartridge screw cap (f), and 2 end caps.
Choose from one of the most recent versions:
Certificates of Analysis (COA)