Skip to Content
Merck
All Photos(1)

Key Documents

2055105

Supelco

Inlet Liner, Split Type, Cup Design (packed with 10% OV-1 on Chromosorb W HP)

pkg of 5 ea

Sign Into View Organizational & Contract Pricing

Select a Size

5 PKG
$501.00

$501.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 PKG
$501.00

About This Item

UNSPSC Code:
12352200

$501.00


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 5 ea

L × O.D. × I.D.

78.5 mm × 6.3 mm × 4.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Agilent (5890, 6890, and 7890)
  • Fits Finnigan (9001GCQ)
  • Fits Varian (CP-1177 Injector)
Inlet liner is an important part within the injection port. The prime functions of inlet liner are to limit sample degradation and improve vaporisation along with correct positioning of syringe needle. A wrong choice of liner can have prominent effect on the chromatogram. The basic problems related with incorrect choice of liner include the higher potential of peak tailing and mass discrimination. The inlet liner volume is recommended to be larger than the amount of sample injected by syringe so as to avoid any reaction with the sample.[1] Injector port liner is suitable to make connection between sample introduction and GC column. Split type has wide bore with 2 or 4 mm I.D. which is suitable for solvent extraction. Cups are used to enable sample mixing.
OV-1 is a modified silicone polymer which has good temperature stability. It is suitable as liquid phases in chromatography. The least polar is a dimethylsilicone which is sold under the mane OV-1.[2]
Chromosorb W, a fragile white solid prepared heating the diatomaceous earth with a flux. The resulting fine structure is cemented over glass which results in large and irregular pores.[3] HP is the classification of a support as high performance grade.[4]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian A. Fowlis
Gas Chromatography, 27-27 (2008)
Robert L. Grob, PhD, Eugene F. Barry, PhD
Modern Practice of Gas Chromatography, 76-76 (2004)
Harold M. McNair, James M. Miller
Basic Gas Chromatography (2011)
Michelle Groves Carlin, John Richard Dean
Forensic Applications of Gas Chromatography, 9-9 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service