Skip to Content
Merck
All Photos(1)

Documents

20048

Supelco

ORBO 601 Amberlite XAD®-8 (16/50), 100 mg (single adsorbent)

W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,W separators

Agency

OSHA 28

composition

Bed A, 100 mg (single-bed sorbent tube)

packaging

pkg of 50 ea

manufacturer/tradename

ORBO 601

O.D. × L

6 mm × 75 mm

matrix

XAD-8

particle size

16-50 mesh

application(s)

environmental
industrial hygiene

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service