19141-U
Hamilton® needles for RN syringes, blunt tip
needle size 26 ga, needle L 51 mm (2 in.), for use with 250μL-10mL syringe
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
blunt tip stainless steel needle (point style 3)
needle size
26 ga
description
blunt tip (stainless steel)
feature
blunt tip (point style 3)
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 7780-02
needle L
51 mm (2 in.)
volume
250-10000 μL
compatibility
for use with 250μL-10mL syringe
for use with Rheodyne Injection Valve
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service