Skip to Content
Merck
All Photos(1)

Key Documents

1895-1AMP

Supelco

F.A.M.E. Mix GLC-40

certified reference material, ampule of 100 mg (Neat)

Synonym(s):

Fatty Acid Methyl Ester Mix

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
$123.00

$123.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 AMP
$123.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

$123.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 100 mg (Neat)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

functional group

carboxylic acid
ester

shipped in

dry ice

storage temp.

-10 to -25°C

General description

This quantitative mix is useful for determining relative retention times and approximating response factors.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methyl arachidate 25 wt. %
    Methyl behenate 25 wt. %
    Methyl palmitate 25 wt. %
    Methyl stearate 25 wt. %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service