Skip to Content
Merck
All Photos(1)

Key Documents

13981-U

Supelco

Metal Packed GC Column (General Configuration)

phase none, matrix 45/60 Molecular Sieve 13X support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
eCl@ss:
32110204
NACRES:
SB.54

material

stainless steel column

parameter

0-400 °C temperature (isothermal or programmed)

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

10.0 ft (3.0 m) × 1/8 in. × 2.1 mm

matrix

45/60 Molecular Sieve 13X support

matrix active group

none phase

column type

packed GC

compatibility

for use with General Configuration

General description

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
Packed column made up of stainless steel are basically used when higher carrier gas flow rates are employed as they can sustain elevated pressure. Stainless steel columns are usually washed with dilute hydrochloric acid, then with water followed by methanol, acetone, methylene dichloride and n-hexane in order to remove any corrosion products and traces of lubricating agents.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Columns for Gas Chromatography: Performance and Selection.
Barry EF and Robert L.
Science, 28-28 (2007)
Introduction to Analytical Gas Chromatography.
Scott PW.
Science, 102-108 (1997)
M Schweigkofler et al.
Journal of hazardous materials, 83(3), 183-196 (2001-05-12)
Methods for the removal of gaseous silicon compounds in biogases are presented. In laboratory studies, various solid adsorption materials and liquid absorption solutions were evaluated for their siloxane elimination efficiencies. Among the liquid sorbents studied, concentrated nitric acid and sulfuric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service