13041-U
Metal Packed GC Column (fits Agilent, Configuration "A")
phase 10% UCW-98, matrix 80/100 Chromosorb P AW support, L × O.D. × I.D. 1.7 ft (0.52 m) × 1/8 in. × 2.1 mm
About This Item
material
stainless steel column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
1.7 ft (0.52 m) × 1/8 in. × 2.1 mm
matrix
80/100 Chromosorb P AW support
matrix active group
10% UCW-98 phase
column type
packed GC
compatibility
for use with Agilent Configuration A
Looking for similar products? Visit Product Comparison Guide
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1 Inhalation
Target Organs
Lungs
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service