11496
PTFE Packed GC Column (General Configuration)
phase none, matrix Chromosil 330 support, L × O.D. × I.D. 8.0 ft (2.4 m) × 1/8 in. × 2.1 mm, Ends empty, middle 6 ft packed
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
8.0 ft (2.4 m) × 1/8 in. × 2.1 mm, Ends empty, middle 6 ft packed
matrix
Chromosil 330 support
matrix active group
none phase
application(s)
pharmaceutical (small molecule)
column type
packed GC
compatibility
for analyte group sulfur compounds
for use with General Configuration
Looking for similar products? Visit Product Comparison Guide
General description
For low temperature applications, a PTFE column offers the flexibility of metal with inertness approaching that of glass. This column is of a general configuration, and can be carefully bent to fit most instruments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service