Skip to Content
Merck
All Photos(11)

Key Documents

ZRB2375

Sigma-Aldrich

Anti-Lyn Antibody, clone 1E23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:2.7.10.2, Lck/Yes-related novel protein tyrosine kinase, Tyrosine-protein kinase Lyn, V-yes-1 Yamaguchi sarcoma viral related oncogene homolog, p53Lyn, p56Lyn

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E23, recombinant monoclonal

description

1E23 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 58.57 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... LYN(4067)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E23 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Lyn. It targets an epitope within 22 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the N-terminal region of human tyrosine protein kinase Lyn A.

Application

Quality Control Testing

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Lyn in RAW264.7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected Lyn in lysates from Ramos cells and 1:1,000 dilution in lysate from Raji cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Lyn in Human tonsil tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound tyrosine protein kinase Lyn peptide with a KD of 2.5 x 10-7 n an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Tyrosine-protein kinase Lyn (UniProt: P07948; also known as EC:2.7.10.2, Lck/Yes-related novel protein tyrosine kinase, V-yes-1 Yamaguchi sarcoma viral related oncogene homolog, p53Lyn, p56Lyn) is encoded by the LYN (also known as JTK8) gene (Gene ID: 4067) in human. Lyn is a non-receptor protein tyrosine kinase of the Src family that plays a role in the regulation of innate and adaptive immune responses, hematopoiesis, responses to growth factors and cytokines, integrin signaling, and responses to DNA damage and genotoxic agents. It is widely expressed and is detected in epidermoid, hematopoietic, and neuronal cells. Its expression is also observed in primary neuroblastoma tumors. Two isoforms of Lyn (Lyn A and Lyn B) have been described that are produced by alternative splicing. They differ by a 20 amino acid region in their SH4 domain that encompasses pY32 motif. Activity of Lyn is regulated by protein interactions through its SH2/SH3 domains as well as via its phosphorylation. In its inactive state, it is phosphorylated at tyrosine 508 by C-terminal Src kinase that creates a binding site for its own SH2 domain. Its SH3 domain can bind an intramolecular proline-motif situated between the SH2 domain (aa 129-226) and kinase domains (aa 247-501), helping generate a stabilized inactive kinase confirmation. Lyn is activated by dephosphorylation of the tyrosine 508 by CD45 and SHP-2 phosphatases. Activation can also occur via interactions with SH2 and/or SH3 domain binding motifs, which compete with Lyn′s own SH3/SH2 intramolecular interaction sites, thereby releasing the auto-inhibitory configuration of the kinase domain. Lyn can then trans-autophosphorylate at tyrosine 397 within the activation loop to generate a highly active enzyme. Lyn plays an important role in the regulation of B-cell differentiation, proliferation, survival, and apoptosis, and is also important for immune self-tolerance. It acts downstream of several immune receptors. Lyn is also reported to down-regulate signaling pathways by phosphorylation of immunoreceptor tyrosine-based inhibitory motifs (ITIM), that then serve as binding sites for phosphatases, such as SHP-1, and SHP-2, that modulate signaling by dephosphorylation of kinases and their substrates. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.(Ref.: Ingley, E. (2012). Cell Commun. Signal. 10; 21; Xu, W., et al. (1999). Mol. Cell. 3(5); 629-638; Hirao, A., et al. (1997). EMBO J. 16(9); 1342-2351; Hata, A., et al. (1994). Mol. Cell Biol. 14(11); 7306-7313).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service