Skip to Content
Merck
All Photos(15)

Key Documents

ZRB2230

Sigma-Aldrich

Anti-VHL Antibody, clone 1C10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Protein G7, pVHL, von Hippel-Lindau disease tumor suppressor

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$350.00
4 X 25 μL
$720.00

$350.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$350.00
4 X 25 μL
$720.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$350.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C10, recombinant monoclonal

description

1C10 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 24.15 kDa
observed mol wt ~17 kDa

purified by

using Protein A

species reactivity

rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... VHL(7428)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C10 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects von Hippel-Lindau disease tumor suppressor (VHL). It targets an epitope within 25 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the internal region of human von Hippel-Lindau disease tumor suppressor (VHL).

Application

Quality Control Testing

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected VHL in Raji cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected VHL in HEK293 and MCF-7 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected VHL in Human colon tissue sections and 1:100 dilution detected VHL in Human testis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound VHL peptide with a KD of 7.7 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected VHL in HEK293 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

von Hippel-Lindau disease tumor suppressor (UniProt: P40337; also known as Protein G7, pVHL) is encoded by the VHL gene (Gene ID: 7428) in human. VHL is a component of the VCB (VHL-Elongin BC-CUL2) complex that acts as a substrate recognition component of a ubiquitin-ligase E3 and directs proteasome-dependent degradation of targeted proteins. VHL interacts with CUL2, and this interaction is dependent on the integrity of the trimeric VCB complex. It targets recruitment subunit in the E3 ubiquitin ligase complex and recruits hydroxylated hypoxia-inducible factor (HIF) under normoxic conditions. It is involved in transcriptional repression through interaction with HIF1α and histone deacetylases. In addition to oxygen-related gene expression, VHL plays a role in cilia formation, cytokine signaling, regulation of senescence, and formation of the extracellular matrix. It is reported to be essential for embryonic and placental development. Its inactivation results in embryonic death from abnormal placental vascularization. Murine models with VHL deficiency develop syncytiotrophoblasts and labyrinths and display hemorrhage by embryonic day 11.5 to 12.5. Three isoforms of VHL have been described that are produced by alternative splicing and alternative initiation. Isoform 1 is predominantly cytoplasmic with some expression in the nucleus and membrane, whereas isoform 3 is equally distributed between the nucleus and cytoplasm and is not observed in the cell membrane. Mutations in VHL gene have been linked to von Hippel-Lindau syndrome, pheochromocytoma, erythrocytosis, and renal cell carcinoma. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Haase, VH. (2009). Curr. Pharm. Des. 15(33); 3895-3903; Kaelin, WG. (2008). Nat. Rev. Cancer. 8; 865-873; Tanimoto, K., et al. (2000). EMBO J. 19(16); 4298-4309).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service