Skip to Content
Merck
All Photos(10)

Documents

ZRB2195

Sigma-Aldrich

Anti-Interferon γ Antibody, clone 1N13 Antibody, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1N13, recombinant monoclonal

description

1N13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 19.35 kDa
observed mol wt ~15 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1N13 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects human Interferon g.

Immunogen

GST/His-tagged recombinant mature form of human Interferon g (without the signal peptide and propeptide).

Application

Quality Control Testing

Evaluated by Western Blotting in NK92 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Interferon in NK92 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected recombinant human Interferon .

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Interferon in human kidney tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Interferon with a KD of 3.3 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 1 µg of a representative lot of this antibody detected interferon-gamma in one million human PBMCs.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Interferon g, clone 1N13, ZooMAb®, Cat. No. ZRB2195, is a recombinant Rabbit monoclonal antibody that specifically targets Interferon g and is tested for use in Affinity Binding Assay, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Interferon g (UniProt: P01579; also known as IFN-gamma, IFN- g , Immune interferon) is encoded by the IFNG gene (Gene ID: 3458) in human. IFN- g is a pleiotropic molecule with anti-proliferative, pro-apoptotic, and anti-tumor properties. It is a type II homodimeric interferon that is synthesized with a signal peptide (aa 1-23) and a propeptide (aa 162-166) that are subsequently cleaved off to produce the mature form. It is secreted predominantly by activated lymphocytes such as CD4 T helper type 1, CD8 cytotoxic T cells, g d T cells, and natural killer cells, and antigen-presenting cells. Its expression is induced by mitogens and cytokines. IFN-g signaling coordinates several biological responses involved in host defense and immune surveillance, establishment of adaptive immunity, and the regulation of inflammation. Its diverse biological activities are initiated by IFN- g -mediated aggregation of at least two different cell surface receptors: IFN gR1 and IFN gR2. IFN- g is reported to activate macrophages toward a pro-inflammatory profile increasing their phagocytic ability and enhancing their microbial killing activity. It is also shown to upregulate cell surface MHC class II on antigen presenting cells, thus promoting peptide-specific activation of CD4 T cells. Mutations in IFNG gene are known to cause aplastic anemia that is characterized by failure of bone marrow to produce adequate number of peripheral blood elements leading to peripheral pancytopenia and hypoplasia. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Castro, F., et al. (2018). Front. Immunol. 9; 847).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service