Skip to Content
Merck
All Photos(7)

Key Documents

ZRB2171

Sigma-Aldrich

Anti-SAFB Antibody, clone 2N15-L2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

HSP27 ERE-TATA-binding protein, HSP27 estrogen response element-TATA box-binding protein, SAF-B, SAF-B1, Scaffold attachment factor B1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$346.00
4 X 25 μL
$711.00

$346.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$346.00
4 X 25 μL
$711.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$346.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2N15-L2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2N15-L2 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Scaffold attachment factor B1 (SAFB). It targets an epitope within 24 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the internal region of human Scaffold attachment factor B1 (SAFB).

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell nuclear extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SAFB in NIH3T3 cell nuclear extract.

Tested Applications

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected SAFB in A431 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected SAFB in Human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Scaffold attachment factor B1 (UniProt: Q15424; also known as SAF-B, SAF-B1, HSP27 estrogen response Element-TATA box-binding protein, HSP27 ERE-TATA-binding protein) is encoded by the SAFB (also known as HAP, HET, SAFB1) gene (Gene ID: 6294) in human. The scaffold attachment factor protein family consists of SAFB1, SAFB2 and a SAF-like transcription modulator (SLTM). SBAF1 is an evolutionarily conserved, monomeric or homodimeric, RNA binding protein that is involved in several  processes, including transcription, chromatin structure, stress response, DNA repair, and apoptosis. It can also form heterodimers with SAFB2. It is ubiquitously expressed with highest levels observed in the central nervous system. Its expression is also observed in breast cancer cell lines. In neurons, its expression is regulated by stress, suggesting its role in coordinating the expression of neuroprotective proteins. It binds to scaffold/matrix attachment region (S/MAR) DNA and forms a molecular assembly point to allow the formation of a transcriptosomal complex for coupling transcription and RNA processing. SAFB1 also interacts with proteins involved in splicing and other aspects of RNA-processing. Four isoforms of SABF1 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hong, EA., et al. (2012). Biochem. Soc. Trans. 40(4); 826-830; Rivers, C., et al. (2015). BMC Biology. 13; Article 111; Nayler, O., et al. (1998). Nucleic Acids Res. 26(15); 3542-3549).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
Element is a trademark of Corning, Inc.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service