Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2096

Sigma-Aldrich

Anti-FosB Antibody, clone 3I8 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

FosB proto-oncogene AP-1 transcription factor subunit, G0/G1 switch regulatory protein 3, Protein FosB, Transcription factor AP-1 subunit FosB

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$346.00
4 X 25 μL
$711.00

$346.00


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$346.00
4 X 25 μL
$711.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$346.00


Estimated to ship on19 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3I8, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... FOSB(2354)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3I8 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects FosB protein. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human FosB.

Application

Quality Control Testing

Evaluated by Western Blotting in HL-60 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected FosB in HL-60 cell lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected FosB in Human placenta tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected FosB in HeLa cells.

Affinity Binding Assay: A representative lot of this antibody bound FosB peptide with a KD of 3.7 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Protein FosB (UniProt: P53539; also known as FosB proto-oncogene AP-1 transcription factor subunit, G0/G1 switch regulatory protein 3, Transcription factor AP-1 subunit FosB) is encoded by the FOSB (also known as G0S3) gene (Gene ID: 2354) in human.  FosB is a member of the Fos family of transcription factors that is shown to accumulate in a subset of neurons of the nucleus accumbens and dorsal striatum following repeated administration of drugs of abuse. It persists in neurons for LONG® periods of  time due to its high stability. The genic expression of FosB seems to be modified after LONG® time exposure to drugs of abuse and these changes may be involved in craving and addicted behavior. The FosB protein exhibits a truncated and stable splicing variant known as ΔFosB, whose accumulation is critical for the establishment of addictive behavior. FosB heterodimerizes with proteins of the Jun family to form an AP-1 transcription factor complex, thereby enhancing their DNA binding activity to gene promoters containing an AP-1 consensus sequence 5′-TGA[GC]TCA-3′. Together with Jun, it also plays a role in activation-induced cell death of T cells by binding to the AP-1 promoter site of Fas ligand (CD95L) and inducing its transcription in response to activation of the TCR/CD3 signaling pathway.  Recurrent rearrangement of Fos and FosB are observed in the most common benign tumors of bone, osteoblastoma, and osteoid osteoma. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Anders, QS., et al. (2018). Front. Pharmacol. 9; 1205; Yin, Z., et al. (2017). Nucleic Acids Res. 45(19); 11425-11436; Baumann, S., et al. (2003). Oncogene. 22(9); 1333-1339; Nestler, EJ., et al. (2001). Proc. Natl. Acad. Sci. USA. 98(20); 11042-11046).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
LONG is a registered trademark of Repligen Sweden AB
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service