Skip to Content
Merck
All Photos(20)

Key Documents

ZRB1567

Sigma-Aldrich

Anti-TLE3 Antibody, clone 1D15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ESG3, Enhancer of split Groucho-like protein 3, Transducin-like enhancer protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D15, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 83.42 kDa
observed mol wt ~80 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TLE3(7090)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1D15 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Transducin-like enhancer protein 3 (TLE3). It targets an epitope within 21 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal region of human Transducin-like enhancer protein 3 (TLE3).

Application

Anti-TLE3, clone 1D15 ZooMAb, Cat. No. ZRB1567, is a recombinant Rabbit monoclonal antibody that specifically targets TLE3 and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TLE3 in HEK293 and NIH3T3 cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TLE3 in HeLa, A431, HepG2, and NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TLE3 in human vaginal tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Transducin-like enhancer protein 3 (UniProt: Q04726; also known as Enhancer of split Groucho-like protein 3, ESG3, TLE3) is encoded by the TLE3 (also known as KIAA1547) gene (Gene ID: 7090) in human. TLE3, a homotetrameric member of the Groucho/TLE family, is a transcriptional corepressor that binds to a number of transcription factors. Groucho/TLE proteins consist of a five-domain structures: a highly conserved Q domain that facilitates oligomerization of Gro/TLE molecules, a glycine/proline-rich (GP) domain that is essential for interaction of these proteins with histone deacetylases, a CcN domain that contains a nuclear localization sequence and CDC22 and casein kinase II phosphorylation sites, a serine/proline-rich domain, and a highly conserved WD40 domain that contains multiple tryptophan and aspartic acid tandem repeats. TLE3 is expressed in white adipose tissue and is shown to enhance the transcriptional activity of peroxisome proliferator-activated receptor (PPAR) and induce adipogenesis in preadipocytes. It also serves as a regulator of MyoD function during myogenesis. It interferes with MyoD function by disrupting the association between the basic helix-loop-helix domain of MyoD and E proteins. TLE3 is also shown to inhibit mitochondrial gene expression in beige adipocytes and conditional deletion of TLE3 in adipocytes is reported to promote mitochondrial oxidative metabolism and increase energy expenditure. It interacts with EBF2 and blocks its ability to promote the thermogenic transcriptional program. TLE3 is ubiquitinated by XIAP/BIRC4, but this ubiquitination does not affect its stability, nuclear localization, or capacity to tetramerize but inhibits its interaction with TCF7L2/TCF4. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Pearson, S., et al. (2019). Genes Dev. 2019. 33(13-14): 747-762; Kokabu, S., et al. (2017). J. Biol. Chem. J. Biol. Chem. 292(31); 12885-12894; Villanueva, CJ et al. (2011). Cell Metab. 13(4); 413-427).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service