Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1505

Sigma-Aldrich

Anti-SIRT6 Antibody, clone 1O6 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Sirtuin-6;NAD-dependent protein deacetylase sirtuin-6;EC:2.3.1.286;Regulatory protein SIR2 homolog 6;SIR2-like protein 6

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$357.00
4 X 25 μL
$735.00

$357.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$357.00
4 X 25 μL
$735.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$357.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O6, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 39.12 kDa
observed mol wt ~40 kDa

purified by

using Protein A

species reactivity

rat, mouse, human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1O6 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects NAD-dependent protein deacetylase sirtuin-6 (SIRT6). It targets an epitope within 15 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human NAD-dependent protein deacetylase sirtuin-6 (SIRT6).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat Brain tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected SIRT6 in Rat Brain tissue lysate.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SIRT6 in A431 and NIH3T3 cells.

Affinity Binding Assay: A representative lot of this antibody bound SIRT6 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

NAD-dependent protein deacetylase sirtuin-6 (UniProt: Q8N6T7; also known as EC:2.3.1.286, Regulatory protein SIR2 homolog 6, SIR2-like protein 6, SIRT6) is encoded by the SIRT6 (also known as SIR2L6) gene (Gene ID: 51548) in human. SIRT6 is a zinc-dependent nuclear enzyme that is involved in various cellular processes, including telomere maintenance, gene expression, genomic stability, cell senescence, and apoptosis. It is associated with telomeric heterochromatin regions in the nucleus. In mitosis, endogenous SIRT6 is shown to localize to the cytoplasm and partially co-localizes with mitotic spindles, and does not associate with condensed chromatin. It is highly expressed in skeletal muscle, heart, liver, and brain where it affects transcriptional regulation in a tissue-specific manner. SIRT6 promotes resistance to DNA damage and oxidative stress, the two principal defects associated with age-related diseases. It has a two-domain structure consisting of a large Rossmann fold and a smaller and structurally varied region that contains the Zinc-binding motif. Its C-terminus is required for its proper nuclear localization, while the N-terminus is important for chromatin association and for its intrinsic catalytic activity. It displays very weak deacetylase activity and can bind NAD+ in the absence of acetylated substrate. It is shown to modulates acetylation of histone H3 in telomeric chromatin during the S-phase of the cell cycle. SIRT6 deficiency results in genomic instability and hypersensitivity to ionizing radiation. Cells deficient in SIRT6 exhibit higher levels of HIF1 activity, increased glucose uptake, and reduced mitochondrial respiration. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Beauharnois JM., et al. (2013). Mol Biosyst. 9(7); 1789-806; Jia, G., et al. (2012). Mol. Cell. Biochem. 364(1-2); 345-350; Ardestani, PM., and Linag, F. (2012). Nucleus. 3(5); 442-451).).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service