Skip to Content
Merck
All Photos(30)

Key Documents

ZRB1458

Sigma-Aldrich

Anti-RAD21 Antibody, clone 2H12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Double-strand-break repair protein rad21 homolog, NXP-1, Nuclear matrix protein 1, SCC1 homolog, hHR21

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$303.00
4 X 25 μL
$625.00

$303.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$303.00
4 X 25 μL
$625.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$303.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H12, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 71.69 kDa
observed mol wt 105 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RAD21(5885)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2H12 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects RAD21. It targets an epitope within 102 amino acids from the C-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 102 amino acids from the C-terminal half of human Double-strand-break repair protein Rad21 homolog (RAD21).

Application

Anti-RAD21, clone 2H12 ZooMAb, Cat. No. ZRB1458, is a recombinant Rabbit monoclonal antibody that specifically targets RAD21 and is tested for used in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RAD21 in NIH3T3 and L6 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected RAD21 in human colon tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RAD21 in A431, HeLa, HUVEC, Jurkat, K562, NIH3T3, and U2OS cells.

Affinity Binding Assay: A representative lot of this antibody bound recombinant RAD21 protein fragment with a KD of 9.0 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Double-strand-break repair protein rad21 homolog (UniProt: O60216; also known as hHR21, Nuclear matrix protein 1, NXP-1, SCC1 homolog) is encoded by the RAD21 (also known as HR21, KIAA0078, NXP1, SCC1) gene (Gene ID: 5885) in human. RAD21 is a double-strand-break repair protein rad21 homolog. It is a component of the cohesion complex and is integral to chromosome segregation, error-free DNA repair, and prevention of inappropriate recombination between repetitive regions. It is cleaved by separase/ESPL1 at the onset of anaphase and this cleavage is required for sister chromatid separation and cytokinesis. The cohesin complex also plays a role in spindle pole assembly during mitosis. RAD21 expression in colorectal cancer is associated with aggressive disease especially in KRAS mutant tumors and resistance to chemoradiotherapy. Mutations in RAD21 gene have been linked to Cornelia de Lange syndrome that is characterized by facial dysmorphisms, abnormal hands and feet, growth delay and cognitive retardation. Some mutations can also cause Mungan syndrome that leads to neuromyopathy, chronic intestinal pseudo-obstruction and a variety of cardiac valve defects. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Deb, S., et al. (2014). Br. J. Cancer 110(6); 1606-1613).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service