Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1384

Sigma-Aldrich

Anti-ALK-7 Antibody, clone 1N5 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$357.00
4 X 25 μL
$735.00

$357.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$357.00
4 X 25 μL
$735.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$357.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1N5, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 54.87 kDa
observed mol wt ~61 kDa

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1N5 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects human Activin receptor-like kinase 7 (ALK-7). It targets an epitope within 17 amino acids from the cytoplasmic domain within the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the cytoplasmic domain within the N-terminal half of human Activin receptor-like kinase 7 (ALK-7), isoform 1.

Application

Quality Control Testing

Evaluated by Western Blotting in Human prostate tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected ALK-7 in Human prostate tissue lysate.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ALK-7 in PC3 cells.

Flow Cytometry Analysis: 1 μg from a representative lot detected ALK-7 in one million PC3 cells.

Affinity Binding Assay: A representative lot of this antibody bound ALK-7 Peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-ALK-7, clone 1N5 ZooMAb®, Cat. No. ZRB1384, is a recombinant Rabbit monoclonal antibody that specifically targets ALK-7 and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, and Western Blotting.

Target description

Activin receptor type-1C (UniProt: Q8NER5, also known as EC:2.7.11.30, Activin receptor type IC, ACTR-IC, Activin receptor-like kinase 7, ALK-7) is encoded by the ACVR1C (also known as ALK7) gene (Gene ID: 130399) in human. ALK-7 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-20) that is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 21-113), a transmembrane domain (aa 114-134, and a cytoplasmic domain (aa 135-493). ALK-7 expression is reported in pancreas, heart, colon, small intestine, ovary and the hippocampus, medulla oblongata and putamen of the brain. It serves as a transmembrane type IC receptor serine kinase for activin or activin-like ligands. The receptor complex consists of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which auto-phosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3. The type I receptors contains a Ser/Thr kinase domain in its cytoplasmic domain (aa 194-485) that is essential for signaling, while the type II are receptors are essential for ligand binding. ALK-7 is reported to negatively regulate glucose-stimulated insulin release in β-cells. Mice lacking ALK-7 display normal pancreas organogenesis but develop an age-dependent syndrome involving progressive hyperinsulinemia, reduced insulin sensitivity, liver steatosis, impaired glucose tolerance, and islet enlargement. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Roberts, HJ., et al. (2003). Biol. Reprod. 68(5); 1719-1726).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the specific amino acid sequence this antibody was generated against?

    1 answer
    1. The exact immunogen sequence is considered proprietary. It consists of 17 amino acids from the cytoplasmic domain (135-493) within the N-terminal half of human Activin receptor-like kinase 7 (ALK-7), isoform 1 (UniProt Q8NER5).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service