Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1322

Sigma-Aldrich

Anti-NPM/B23 Antibody, clone 2H21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Nucleophosmin;Nucleolar phosphoprotein B23;Nucleolar protein NO38;Numatrin)

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$350.00
4 X 25 μL
$720.00

$350.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$350.00
4 X 25 μL
$720.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$350.00


Estimated to ship on09 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H21, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 32.58 kDa
observed mol wt ~36 kDa

purified by

using Protein A

species reactivity

rat, human

species reactivity (predicted by homology)

bovine, monkey, porcine, canine, feline, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NPM1(4869)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2H21 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects NPM/B23 (Nucleophosmin). It targets an epitope within 20 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human.

Application

Quality Control Testing

Evaluated by Western Blotting in L6 cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected NPM/B23 in L6 cell lysates.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected NPM/B23 in U2OS cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected NPM/B23 in U2OS cells.

Affinity Binding Assay: A representative lot of this antibody bound NPM/B23 peptide with a KD of 5.6 x 10-7 in an affinity binding assay.

Enzyme Immunoassay (ELISA) Analysis: Various dilutions from a representative lot detected His-tagged, full-length, recombinant Human B23-NPM1 protein.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected NPM/B23 in human colon tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Nucleophosmin (UniProt: P06748; also known as NPM, Nucleolar phosphoprotein B23, Nucleolar protein NO38, Numatrin, NPM) is encoded by the NPM1 (also known as NPM) gene (Gene ID: 4869) in human. NPM is a member a histone chaperone family that is abundant in nucleus of proliferating cells. It participates in ribosome biogenesis, mRNA processing, chromatin remodeling, embryogenesis, and regulation of tumor suppressors p53/TP53. It is generally found in the nucleolus but is translocated to the nucleoplasm following serum starvation or treatment with anticancer drugs. It is also detected in cytoplasm of subjects with primary acute myelogenous leukemia (AML), but not with secondary AML. It is reported to bind ribosome presumably to drive ribosome nuclear export. It acts as a chaperonin for the core histones H3, H2B and H4. NPM also stimulates APEX1 endonuclease activity on apurinic/apyrimidinic double-stranded DNA but inhibits APEX1 endonuclease activity on AP single-stranded RNA. It may also exert a control of APEX1 endonuclease activity within nucleoli devoted to repair AP on rDNA and the removal of oxidized rRNA molecules. It is phosphorylated on serine 4 by PLK1 and PLK2. Phosphorylation by PLK1 takes placed during mitosis and phosphorylation by PLK2 is required for centriole duplication and is sufficient to trigger centriole replication. It is also phosphorylated at serine 125 and threonine 199 by Cdk2. Phosphorylation at threonine 199 increases its affinity for ROCK2 and also triggers initiation of centrosome duplication. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Box, JK., et al. (2016). BMC Mol. Biol. 17(1); Article 19; Krause, A., and Hoffmann, I. (2010). PLoS One. 5(3); e9849; Swaminathan, V., et al (2005). Mol. Cell Biol. 25(17); 7534-7545).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.03 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service