Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1275

Sigma-Aldrich

Anti-CD81 Antibody, clone 1C23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$303.00
4 X 25 μL
$625.00

$303.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$303.00
4 X 25 μL
$625.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$303.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C23, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~25.81 kDa
observed mol wt ~19 kDa

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C23 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects CD81. It targets an epitope within 23 amino acids from the first extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the first extracellular domain of human CD81.

Application

Quality Control Testing

Evaluated by Western Blotting in Human Testis tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD81 in Human Testis tissue lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CD81 in human tonsil tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound CD81 peptides with a KD of 9.8 x 10-8 in an affinity binding assay.

Flow Cytometry Analysis: 1 μg from a representative lot detected CD81 in one million MOLT4 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CD81, clone 1C23 ZooMAb®, Cat. No. ZRB1275, is a recombinant Rabbit monoclonal antibody that specifically detects CD81 and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

CD81 antigen (UniProt: P60033; also known as 26 kDa cell surface protein TAPA-1, Target of the antiproliferative antibody 1, Tetraspanin-28, Tspan-28, CD81) is encoded by the CD81 (also known as TAPA1, TSPAN28) gene (Gene ID: 975) in human. TAPA-1 is a nonglycosylated, homodimeric, multi-pass membrane protein of the tetraspanin (TM4SF) family. It is expressed in a wide range of cells, including hematolymphoid, neuroectodermal, and mesenchymal tumor cell lines and is reported to play a role in the regulation of lymphoma cell growth. It contains four transmembrane domains, two extracellular loops, and short amino and carboxyl termini that are intracellular. TAPA-1 is shown to affect cell adhesion, morphology, activation, proliferation, and differentiation of B, T, and other cells. On B cells it is a part of a complex involving CD21, CD19, and Leu13 and this complex is reported to reduce the threshold for B cell activation via the B cell receptor by bridging antigen specific recognition and CD21-mediated complement recognition. On T cells, it is shown to associate with CD4 and CD8 and provides a costimulatory signal with CD3. It is also reported to act as a receptor for hepatitis C virus (HCV) in hepatocytes. Mutations in CD81 gene cause common variable immunodeficiency 6 (CVID6) that is characterized by antibody deficiency, hypogammaglobulinemia, recurrent bacterial infections, and an inability to mount an antibody response to antigen. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Levy S., et al. (1998). Annu. Rev. Immunol.16; 89-109; Oren, R., et al. (1990). Mol. Cell Biol. 10(8); 4007-15).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service