Skip to Content
Merck
All Photos(21)

Key Documents

ZRB1213

Sigma-Aldrich

Anti-phos-AMPKα (Thr172) Antibody, clone 5J23 , ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

5′-AMP-activated protein kinase catalytic subunit alpha-2, ACACA kinase, AMPK subunit alpha-2, Acetyl-CoA carboxylase kinase, EC: 2.7.11.1, HMGCR kinase, Hydroxymethylglutaryl-CoA reductase kinase

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$357.00
4 X 25 μL
$735.00

$357.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$357.00
4 X 25 μL
$735.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$357.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5J23, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 62.32 kDa
observed mol wt ~62 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pThr172)

Gene Information

human ... PRKAA2(5563)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 5J23 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects AMPK alpha2. It targets an epitope within 10 amino acids surrounding phosphothreonine 172.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphothreonine 172 from human AMPK alpha 2.

Application

Anti-phospho-AMPK (Thr172), clone 5J23 ZooMAb, Cat. No. ZRB1213, is a specific recombinant rabbit monoclonal antibody that detects AMPK alpha phosphorylated on threonine 172 and has been tested for use in Immunocytochemistry, Immunohistochemistry, Peptide Inhibition Assay, and Western Blotting.
Peptide Inhibition Analysis: Preincubation of a 1:1000 dilution from a representative lot with phosphor-AMPK (Thr172) immunogen peptide abolished target band detection in lysate from MCF7 cells treated with Oligomycin (0.5 uM, 30 min), but not in the corresponding nonphosphopeptide.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected phospho-AMPK (Thr172) in human skin and human testis tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected phospho-AMPK (Thr172) in HeLa, A431, HUVEC, and NIH 3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens and experimental conditions may vary.

Target description

5′-AMP-activated protein kinase catalytic subunit alpha-2 (UniProt: P54646; also known as EC: 2.7.11.1, AMPK subunit alpha-2, Acetyl-CoA carboxylase kinase, ACACA kinase, Hydroxymethylglutaryl-CoA reductase kinase, HMGCR kinase) is encoded by the PRKAA2 (also known as AMPK, AMPK2) gene (Gene ID: 5563) in human. AMPK is a heterotrimer composed of an alpha catalytic subunit (PRKAA1 or PRKAA2), a beta (PRKAB1 or PRKAB2) and a gamma non-catalytic subunit (PRKAG1, PRKAG2 or PRKAG3). It also contains an autoinhibitory sequence (aa 291-376) that shows some sequence similarity with the ubiquitin-associated domains and it represses kinase activity. Its protein kinase domains is localized to amino acids 16-268. AMPK is an energy sensor protein kinase that plays a key role in regulating cellular energy metabolism. In response to reduction of intracellular ATP levels, AMPK activates energy-producing pathways and inhibits energy-consuming processes, including inhibition of protein, carbohydrate and lipid biosynthesis, and reduces cell growth and proliferation. AMPK acts via direct phosphorylation of metabolic enzymes, and by longer-term effects via phosphorylation of transcription regulators. AMPK is shown to regulate insulin-signaling and glycolysis by phosphorylating IRS1, PFKFB2 and PFKFB3 and it stimulates glucose uptake in muscle by increasing the translocation of the glucose transporter GLUT4 to the plasma membrane. In the liver it acts as a key regulator of glucose homeostasis by phosphorylating CRTC2/TORC2, leading to CRTC2/TORC2 sequestration in the cytoplasm. AMPK is activated by phosphorylation on threonine 172 by STK11/LKB1 or by CAMKK2 triggered by increase in intracellular calcium. Binding of AMP to non-catalytic gamma subunit results in allosteric activation, inducing phosphorylation on this site. AMP-binding to gamma subunit also sustains activity by preventing dephosphorylation of threonine 172. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service