Skip to Content
Merck
All Photos(13)

Key Documents

ZRB1165

Sigma-Aldrich

Anti-Myosin-9/MYH9 Antibody, clone 3O13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cellular myosin heavy chain type A;Myosin heavy chain 9;Myosin heavy chain non-muscle IIa;Non-muscle myosin heavy chain A;NMMHC-A;Non-muscle myosin heavy chain IIa;NMMHC II-a;NMMHC-IIA

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$357.00
4 X 25 μL
$735.00

$357.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$357.00
4 X 25 μL
$735.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$357.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3O13, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 226.53 kDa

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG

epitope sequence

C-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MYH9(4627)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3O13 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Mysosin-9 (MYH9). It targets an epitope within 16 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human Myosin-9 (MYH9).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected Myosin-9/MYH9 in Human kidney tissue sections.

Tested Applications

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Myosin-9/MYH9 in C2C12 cells.

Affinity Binding Assay: A representative lot of this antibody bound Myosin-9 peptide with a KD of 2.1 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Myosin-9 (UniProt: P35579; also known as Cellular myosin heavy chain type A, Myosin heavy chain 9, Myosin heavy chain non-muscle IIa, Non-muscle myosin heavy chain A, NMMHC-A, Non-muscle myosin heavy chain IIa, NMMHC II-a, NMMHC-IIA) is encoded by the MYH9 gene (Gene ID: 4627) in human. Myosins are highly conserved, ubiquitously expressed proteins that form regular bipolar thick filaments that together with actin thin filaments constitute the fundamental contractile unit of skeletal and cardiac muscle. Along with actin they serve as motor molecules with ATPase activity that is essential for muscle contraction. Myosin consists of 2 heavy chain subunits (MHC), 2 light chain subunits (MLC) and 2 regulatory light chain subunits (MLC-2). MLC-2C plays an important role in regulation of both smooth muscle and non-muscle cell contractile activity via its phosphorylation. Myosin-9 is a cellular myosin that is expressed in kidney and leukocytes. Its actin-binding region is localized in amino acids 654-676. Its rod-like tail sequence is highly repetitive, showing cycles of a 28-residue repeat pattern composed of 4 heptapeptides that is characteristic for -helical coiled coils. Myosin-9 plays a role in cytokinesis, cell shape, secretion, and capping. It is also required for cortical actin clearance prior to oocyte exocytosis. It promotes cell motility in conjunction with A100A4 protein. During cell spreading, myosin-9 is reported to play a role in cytoskeleton reorganization, focal contact formation, and lamellipodial retraction. Genetic variations in MYH9 have been linked with non-diabetic end stage renal disease. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Pazik, J., et al. (2020). BMC Nephrol. 21; Article 380; Li, Z-H., and Bresnick, AR. (2006). Cancer Res. 66(10); 5173-5180).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service