Skip to Content
Merck
All Photos(8)

Key Documents

ZMS1120

Sigma-Aldrich

Anti-Laminin α2 80 kDa Antibody, clone 5H2 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Laminin M chain, Laminin-12 subunit alpha, Laminin-2 subunit alpha, Laminin-4 subunit alpha, Merosin heavy chain

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$346.00
4 X 25 μL
$711.00

$346.00


Estimated to ship on11 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$346.00
4 X 25 μL
$711.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$346.00


Estimated to ship on11 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5H2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

rabbit, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 5H2 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects Laminin 2. It targets an epitope within the C-terminal region.

Immunogen

80 kDa fragment from the C-terminal region of human Laminin 2.

Application

Quality Control Testing

Evaluated by Western Blotting in Human heart tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Laminin 2 in Human Heart tissue lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Laminin 2 in human heart and human cerebral cortex tissue sections.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected Laminin 2 in human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Laminin subunit alpha-2 (UniProt: P24043; also known as Laminin M chain, Laminin-12 subunit alpha, Laminin-2 subunit alpha, Laminin-4 subunit alpha, Merosin heavy chain) is encoded by the LAMA2 (also known as LAMM) gene (Gene ID: 3908) in human. Laminins are a family of heterotrimeric extracellular glycoproteins consisting of α, β, and γ chains that are bound to each other by disulfide bonds into a cross-shaped molecule comprising one LONG® and three short arms with globules at each end.  Thus far five α, three β, and three γ subunits are known, which assemble into at least 15 different laminin molecules.  Laminin α2 is a glycoprotein with multiple EGF-like domains and is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off. It is expressed in multiple tissues, including placenta, striated muscle, cardiac muscle, pancreas, lung, spleen, skin, bone, and some regions of the brain. Laminin α2 binds to cells via a high Affinity receptor and mediates the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components.  Mutations in LAMA2 gene have been linked to Merosin-deficient congenital muscular dystrophy 1A (MDC1A) and limb-girdle muscular dystrophy that results in muscle weakness and atrophy.  This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zhang, X., et al. (1996). J. Biol. Chem. 271(44); 27664-27669; Ehrig, K., et al., (1990). Proc. Natl. Acad. Sci. USA. 87(9); 3264-3268).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
LONG is a registered trademark of Repligen Sweden AB
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service