Skip to Content
Merck
All Photos(1)

Key Documents

ZFNDHFR

SAFC

CompoZr® Knockout ZFN Kit- CHO DHFR

dihydrofolate reductase

Synonym(s):

CompoZr® Knockout ZFN Kit for CHO Cells

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$29,882.00

$29,882.00


Check Cart for Availability


Select a Size

Change View
50 μG
$29,882.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.75

$29,882.00


Check Cart for Availability

Quality Level

product line

CompoZr®

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

CompoZr Knockout ZFNs create targeted double strand breaks at a gene specific target locus. Through the cellular process of Non-Homologous End Joining, this double strand break can result in modification of the DNA sequence and therefor create a functional knockout of the targeted gene. To provide the best chance of creating a functional gene knockout, CompoZr Knockout ZFNs are targeted to the first 2/3 of the coding region for the gene of interest.

Physical form

CompoZr Knockout ZFNs are provided as two plasmids encoding for the gene specific zinc finger nuclease.

Legal Information

All Zinc Finger Nucleases are sold under license from Sangamo Biosciences. For a copy of the Label License provided with purchase of CompoZr ZFNs, please visit the ZFN Label License page.
CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In the present study, we have identifi ed species-specifi c housekeeping genes (HKGs) for Chinese Hamster Ovary (CHO) cells using data from microarray gene expression profiling.

MGAT1 adds N-acetylglucosamine to the Man5GlcNAc2 (Man5) structure. Goh et al. reported increased sialylation after restoring MGAT1 function in MGAT1 deficient CHO cells.

A signal peptide is a 5-30 amino acid (aa) peptide present at the N-terminus of secretory proteins.

Related Content

Learn more about cell line development and the relevant products you need for this process.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service