Key Documents
Z760986
PCV cell counting tubes
capillary graduations only, no cap
Synonym(s):
PCV Tubes
Select a Size
50 EA
$332.00
$332.00
Check Cart for Availability
Select a Size
Change View
50 EA
$332.00
About This Item
UNSPSC Code:
41121706
NACRES:
NB.22
$332.00
Check Cart for Availability
material
(PCV)
colorless tube
conical bottom
straight neck
sterility
non-sterile
packaging
pack of 50 ea
manufacturer/tradename
TPP 87005
availability
not available in (available only in North America)
General description
The packed cell volume (PCV) tube is a quick and reproducible measuring technique of the cell volume that is suitable to monitor cell growth (cell mass).
Features and Benefits
  • Present with or without scale.
  • Fits in microcentrifuges.
  • Caps available if required.
  • Ideal to monitor cell growth
recommended
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Yan Xu et al.
Journal of nanobiotechnology, 15(1), 50-50 (2017-07-12)
Increasing production of nanomaterials requires fast and proper assessment of its potential toxicity. Therefore, there is a need to develop new assays that can be performed in vitro, be cost effective, and allow faster screening of engineered nanomaterials (ENMs). Herein
Xiqian Jiang et al.
ACS chemical biology, 10(3), 864-874 (2014-12-23)
Glutathione (GSH) plays an important role in maintaining redox homeostasis inside cells. Currently, there are no methods available to quantitatively assess the GSH concentration in live cells. Live cell fluorescence imaging revolutionized the field of cell biology and has become