Skip to Content
Merck
All Photos(1)

Key Documents

Z744278

Wilmad® Suprasil® VT Dewar Inserts for Bruker®

OAL, O.D. 10 mm , lower, L 147 mm , upper, L 72 mm , lower

Synonym(s):

EPR cryo dewar, EPR dewar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103800
NACRES:
NB.15
Pricing and availability is not currently available.

packaging

pack of 1 ea

manufacturer/tradename

Wilmad WG-821-STW-AH-Q

L

147 mm , upper
72 mm , lower

O.D.

10 mm , lower

Looking for similar products? Visit Product Comparison Guide

General description

Bruker style Variable Temperature Dewar Inserts are not silvered, and do not offer a dimple to support the sample tube. Maximum working temperature is 600 K.Dimension A: 147mm, Dimension B: 72mm, Dimension C: O.D. = 10.5mm, Dimension D: I.D. = 6mm

Application

EPR

Features and Benefits

Maximum working temperature is 600 K

Legal Information

Bruker is a registered trademark of Bruker-Physik AG
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service