Skip to Content
Merck
All Photos(1)

Key Documents

Z744114

Freezer box divider

to hold, 81 tube(s), pack of 12

Synonym(s):

freezer container, freezer storage, storage container

Sign Into View Organizational & Contract Pricing

Select a Size

12 EACH
$37.80

$37.80


Check Cart for Availability


Select a Size

Change View
12 EACH
$37.80

About This Item

UNSPSC Code:
41122804

$37.80


Check Cart for Availability

feature

to hold 81 tube(s)

packaging

pack of 12

manufacturer/tradename

Diversified Biotech FBCD-2000

Looking for similar products? Visit Product Comparison Guide

General description

Freezer box divider, 81 hole 12/pk

Application

Cryogenics

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service