Key Documents
Z743323
Breathe-EASY® tube membranes for microtubes, , polyurethane, precut sizes
sterile
Synonym(s):
breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture
Select a Size
100 EA
$69.20
$69.20
Estimated to ship on23 May 2025
Select a Size
Change View
100 EA
$69.20
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
$69.20
Estimated to ship on23 May 2025
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Diversified Biotech BMTM-1000
General description
Breathe-EASY polymer-type membrane offers a water vapor transmission rate of 0.7 kg/ m2/24 hours.
  • Membranes are ideal for cell growth, concentrating samples (e.g. freeze drying), and gas exchange.
  • Easy microtube cap closure. Will not bind in centrifuge rotors.
  • Formatted with an easy-to-use finger tab. Pre-cut to fit a variety of popular tube sizes. Convenient scoring makes liner removal quick and easy.
  • Membranes are gamma irradiated to ensure sterility. Free of cytotoxins.
Legal Information
Breathe-Easy is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1