Key Documents
Z742662
Tube-O-Tank for use with the Tube-O-DIALYZER
small
Select a Size
1 EA
$286.00
$286.00
Estimated to ship on21 May 2025
Select a Size
Change View
1 EA
$286.00
About This Item
UNSPSC Code:
41123100
NACRES:
NB.24
$286.00
Estimated to ship on21 May 2025
packaging
pkg of 1 ea
manufacturer/tradename
G-BioSciences 786-145D
General description
Dialysis tank specifically designed for use with the Tube-O-DIALYZER. Two sizes are available that are suitable for Micro (Small) and Medi (Large) size Tube-O-DIALYZER Each tank will accommodate 7 Tube-O-DIALYZER and are supplied with floats.
Legal Information
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)