Skip to Content
Merck
All Photos(1)

Key Documents

Z741771

Sigma-Aldrich

Sigma Aldrich® Dual Run & Blot Accessory

Buffer Dam

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$47.10

$47.10


Check Cart for Availability


Select a Size

Change View
1 EA
$47.10

About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

$47.10


Check Cart for Availability

material

white plastic plate

manufacturer/tradename

Sigma-Aldrich

size

10 cm × 10 cm

Looking for similar products? Visit Product Comparison Guide

General description

Sigma Aldrich gels run best on our Dual Run and Blot unit. This unique apparatus permits the running of both 10x10cm and 8x10cm gels. The unit is available in dual and quad form enabling up to 4 gels to be run at one time.

Both units can also be used to complete Western blots. The unique electrode arrangement provides a gradient electric field which ensures that both small and large proteins migrate from the gel at a comparable rate.

Application

A Sigma Aldrich Buffer dam is used in place of a gel when only one gel is to be run. Simple insert into the Dual Run & Blot Core Module as if it were a Sigma Aldrich Precast Gel Cassette.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service