Skip to Content
Merck
All Photos(1)

Key Documents

Z741696

Nalgene® syringe filter units, disposable, polyethersulfone (PES) membrane

pore size 0.2 μm, diam. 13 mm

Synonym(s):

sterile filtration, syringe filters

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$718.00

$718.00


Available to ship on16 April 2025Details



Select a Size

Change View
100 EA
$718.00

About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

$718.00


Available to ship on16 April 2025Details


material

polyethersulfone membrane
polypropylene housing (w/green ring)

sterility

sterile (Certified sterile, non-cytotoxic and non-pyrogenic)

packaging

pkg of 100 ea

manufacturer/tradename

Nalgene® 720-1320

volume measuring range

2-10 mL

diam.

13 mm

pore size

0.2 μm

fitting

female inlet (Luer-Lok®)
male outlet (Luer slip)

suitability

suitable for application (sterile filtration and clarification of cell culture media, buffers and additives)

Looking for similar products? Visit Product Comparison Guide

General description

The universal tissue culture membrane (PES), offers higher flow rates, lower protein binding and lower extractables than competitive cellulosic membranes. Inherently hydrophilic; contains no external wetting agents.

  • Sample volume size: 2-10 mL
  • Inlet: Female Luer-Lok®
  • Outlet: Male Luer slip

Certified sterile, non-cytotoxic and non-pyrogenic. Do not autoclave.

Application

Recommended for cell/tissue culture applications.

Legal Information

Luer-Lok is a registered trademark of Becton-Dickinson & Co.
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is the filter hydrophilic?

    1 answer
    1. Yes, the filter membrane is hydrophilic.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service