Key Documents
Z741608
Plus Safe-Cone Filter
diam. 6.73 mm, polyethylene
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Product Z741608 is not currently sold in your country. Contact Technical Service
material
polyethylene
packaging
package of 50 (individually wrapped)
manufacturer/tradename
Sartorius 721015
diam.
6.73 mm
General description
The unique and replaceable polyethylene (PE) Safe-Cone Filters act as a final barrier to prevent any fluids and liquid vapours from reaching the internal components of the pipette.Standard Safe-Cone Filters are for general applications. They can be used for the same type of work as the Standard Filter is recommended for, but need to be changed less frequently. Self-sealing Plus Safe-Cone Filters are meant for applications such as radioactive work, cell culture, bacterial and virological work and molecular biology.The interval of changing the filter depends on the application and the sample. However, according to studies, the filter should be changed daily (or after 50 to 250 pipettings) and immediately in case of over-aspiration. To ensure safety of the user, forceps should be used to change the filter. The mLINEmLINE® pipettes also feature a built-in filter ejector. In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter.The unique and replaceable polyethylene (PE) Safe-Cone Filters act as a final barrier to prevent any fluids and liquid vapours from reaching the internal components of the pipette.Standard Safe-Cone Filters are for general applications. They can be used for the same type of work as the Standard Filter is recommended for, but need to be changed less frequently. Self-sealing Plus Safe-Cone Filters are meant for applications such as radioactive work, cell culture, bacterial and virological work and molecular biology.The interval of changing the filter depends on the application and the sample. However, according to studies, the filter should be changed daily (or after 50 to 250 pipettings) and immediately in case of over-aspiration. To ensure safety of the user, forceps should be used to change the filter. The mLINE® pipettes also feature a built-in filter ejector. In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter.
Legal Information
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)