Z740473
MICROGARD™ 2000 coveralls for protection from infective agent
size 2XL (elasticated ankles)
Select a Size
$25.90
Select a Size
About This Item
$25.90
Recommended Products
size
2XL (elasticated ankles)
manufacturer/tradename
Arco 1762007
General description
Features:
- 3-piece hood
- 2-way front zipper with re-sealable storm flap
- Finger loops
- Elasticated hood, waist, wrists and ankles
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-5 Anti-static
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)