Skip to Content
Merck
All Photos(1)

Key Documents

Z724408

Hettich® adapter for rotors 1013, 1015, 1115, 1416, 1613 and 1615

for EBA 21, MIKRO 220R, ROTOFIX 32A and UNIVERSAL 320/320R Centrifuges, Adapter for various tubes/rotors

Synonym(s):

Hettich® centrifuge adapters

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$78.60

$78.60


Estimated to ship on01 June 2025



Select a Size

Change View
1 EA
$78.60

About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

$78.60


Estimated to ship on01 June 2025


feature

Holds 5 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Hettich 1054-A

technique(s)

centrifugation: suitable

compatibility

for use with 1015, 1139, 1613, 1615 (rotors)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Compatible with Rotors 1013, 1015 (Z722758), 1115 (Z601217), 1416 (Z722766), 1613 and 1615 (Z724297)

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service