Z724173
Hettich® MIKRO 220/220R centrifuge
MIKRO 220R, AC/DC input 240 V AC
Synonym(s):
Hettich MIKRO 220
About This Item
Recommended Products
AC/DC input
, Schuko plug
, UK plug
feature
CE compliant
cooling
refrigerated
manufacturer/tradename
Hettich 2205
parameter
temp. range at -20-40 °C
18000 rpm
31514 rpm
technique(s)
centrifugation: suitable
H × W × D
313 mm (12.5 in.) × 330 mm (13 in.) × 650 mm (25.5 in.)
weight
42 kg (92.5 lb)
compatibility
CSA or cTUVus label for Canada (SCC)
Looking for similar products? Visit Product Comparison Guide
General description
These centrifuges are used in clinical laboratories as well as in genetic research, virology, cytology and bacteriology. Capacity, performance and cooling of the MIKRO 220R specifically designed to meet the demands of sample preparation for PCR applications.
Rotors and other centrifuge accessories are sold separately.
- Available in non-refrigerated (220) and refrigerated (220R) versions
- Accomodates 0.2-2.0 mL microliter tubes (The refrigerated MIKRO 220R can also accomodate tubes up to 50 mL)
- Extremely short run-up and run-down times even with rotors attaining higher speeds
- Centrifuge chamber of stainless steel
- Powered lid-locking with viewing port in lid
- Quick entry foil keypad
- Lid locking and holding during rotor run
- Lid dropping protection
- Emergency lid-lock release
- Motor-overheating protection
- Chamber overheating protection (MIKRO 220R only)
- Imbalance switch-off
- Automatic rotor recognition
- Easily exchangeable rotors
- 10 programmable memories
Rotors and other centrifuge accessories are sold separately.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service