Key Documents
Z722790
Hettich® ROTOFIX 32A/Universal 320/320R centrifuge accessories
Rotor, 4 place, for ROTOFIX 32A and UNIVERSAL 320
Select a Size
1 EA
$2,740.00
$2,740.00
Check Cart for Availability
Select a Size
Change View
1 EA
$2,740.00
About This Item
UNSPSC Code:
41103900
NACRES:
NB.82
$2,740.00
Check Cart for Availability
feature
to hold 4 x 1389, 1490 buckets
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1324
parameter
3,328 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Rotafix 32A and Universal 320/320R
Related Categories
General description
Carriers not included.
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)