Skip to Content
Merck
All Photos(1)

Documents

Z720364

Ma-GEL-in Western Transfer Protein System

gel size 10 cm × 10 cm, volt limit: 100 vdc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NC.01

gel size

10 cm × 10 cm

General description

The Western Transfer Protein System allows rapid transfer of protein/peptide samples from Polyacrylamide gels to membranes of either 8 x 10 cm or 10 x 10 cm formatted gels. The transfer system accommodates two cassettes for transfer depending upon the format. Mini PAGE gels or Mini Agarose gels can be transferred in the module within a high intensity field while maintaining comparable resolution. The IB95000 fits directly into theIB92000 buffer tank.
  • Interior Transfer Reservoir constructed of 3/8 in. acrylic
  • Interior reservoir overall dimensions: 14 cm(L) x 6.5 cm(W) x 9 cm(H)
  • Electrodes: 30ga. platinum wire
  • Maximum gel size: 10 cm x 10 cm
  • Voltage limit: 100 vdc
  • Current limit: 500 ma
  • Unit comes with two transfer cassettes w/locks and four foam pads

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service