Skip to Content
Merck
All Photos(1)

Key Documents

Z709115

ART® self-sealing barrier pipette tips

ART Softfit 50U, volume range 0.5-50 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$413.00
5 PKG
$1,530.00

$413.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$413.00
5 PKG
$1,530.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
0.5—50 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2780

$413.00


Check Cart for Availability

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

pack of 960 ea (10 racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2780

technique(s)

PCR: suitable

maximum volume

50 μL

volume range

0.5-50 μL

volume

0.5—50 μL

compatibility

for use with

  • Biohit Electronic (Limited Draw) 5-100 μl
  • Biohit Variable 5-50 μl
  • Biohit Multi-Channel 5-50 μl
  • Capp Aero 50 μl
  • Eppendorf Research Pro 4860 5-100 μl
  • Eppendorf Titerman 4908 5-50 μl
  • Finnpipette Multi-Channel 5-50 μl
  • Rainin EDP-Series 100 (Limited Draw) 10-100 μl
  • Socorex/Wheaton 822 Series (Limited Draw) 5-100 μl

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Universal SoftFit-L design to fit most 50 μl multi-channel pipettors

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).[1][2][3]

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service