Key Documents
Z709077
ART® non-filtered tip
HLT 300U SOFTFIT, size 1-300 μL, Pure packed, non-sterile
Synonym(s):
pipet tips, pipette tips
Select a Size
1 PKG
$347.00
5 PKG
$1,290.00
$347.00
Check Cart for Availability
Select a Size
Change View
1 PKG
$347.00
5 PKG
$1,290.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
1—300 μL
sterility:
non-sterile
feature:
DNase free
RNase free
Pure packed
packaging:
pack of 960 ea
manufacturer/tradename:
Molecular BioProducts 3771
$347.00
Check Cart for Availability
sterility
non-sterile
feature
DNase free
RNase free
Pure packed
packaging
pack of 960 ea
manufacturer/tradename
Molecular BioProducts 3771
maximum volume
300 μL
size
1-300 μL
volume
1—300 μL
Related Categories
General description
Easily access pipette tips with one hand using the user-friendly ART® non-filtered tips. It is designed to match your workflow and avoid concerns about tip spills on your bench or floor.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Features and Benefits
  • Ensures quality, performance, and reliability.
  • Certified sterile tips are free of DNase, RNase, DNA, and pyrogen contamination.
  • Racks designed for convenient and efficient access to pipette tips, saving valuable time during experiments.
  • Front latch mechanism securely closes and locks, providing safe and protected storage of tips.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)