Skip to Content
Merck
All Photos(1)

Key Documents

Z707384

SealPlate® MiniStrips film

non-sterile

Sign Into View Organizational & Contract Pricing

Select a Size

200 EA
$77.90

$77.90


Estimated to ship on29 May 2025



Select a Size

Change View
200 EA
$77.90

About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

$77.90


Estimated to ship on29 May 2025


material

clear polyester film

sterility

non-sterile

feature

piercable

packaging

pack of 200 ea

manufacturer/tradename

Excel Scientific SP-2X8-50

technique(s)

ELISA: suitable

L

102.9 mm

W × L

1.9 cm × 10.29 cm , 8.26 cm with Tabs removed

W

82.6 mm

Looking for similar products? Visit Product Comparison Guide

General description

MiniStrips films are identical to SealPlate films, but designed for sealing one or two 8 well rows at a time. They may be used on strip-well plates or standard plates whenever rows must be selectively protected or accessed. Use as a secondary seal to cover films that have already been pierced.

Legal Information

MiniStrips is a trademark of Excel Scientific, Inc.
SealPlate is a registered trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this product DNAse/RNAse free?

    1 answer
    1. This product is tested for suitability in Elisa and has not been tested for the presence of DNase or RNase. Please see the link below to review the PCR-suitable FilmStrips, which are RNase and DNase-free:
      https://www.sigmaaldrich.com/product/sigma/z744065

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service