Skip to Content
Merck
All Photos(2)

Key Documents

Z694592

Nanofiber cell culture dish, with aligned nanofibers

diam. 100 mm

Synonym(s):

3d plates, tissue culture plates

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$72.10

$72.10


Check Cart for Availability


Select a Size

Change View
1 EA
$72.10

About This Item

UNSPSC Code:
41121812
NACRES:
NB.22

$72.10


Check Cart for Availability

material

round

sterility

sterile

feature

lid: no

packaging

pack of 1 ea

manufacturer/tradename

Nanofiber Solutions 10002

diam.

100 mm

suitability

suitable for (Tissue Engineering, Cancer Research)

binding type

(plasma surface treated)

Looking for similar products? Visit Product Comparison Guide

General description

Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for Cancer Research. The Cells can be simply removed for protein or gene analysis.

The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.

The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service