Z688843
Nunc® UpCell™ Surface cell culture dish
size 60 mm, w/ grid, sterile, case of 30 ea
Synonym(s):
cell culture dish, tissue culture dish
Select a Size
$2,230.00
Estimated to ship on28 May 2025
Select a Size
About This Item
$2,230.00
Estimated to ship on28 May 2025
Recommended Products
material
polystyrene
sterility
sterile
feature
lid vented
membrane not included
w/ grid
packaging
case of 30 ea
manufacturer/tradename
Nunc 174906
dish diam. × H
60 mm × 15 mm
external L × W
60 mm × 15 mm
size
60 mm
surface area
21.5 cm2 , culture area (cm2)/well
working volume
5 mL
binding type
(UpCell Surface)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Harvesting of cells with intact surface proteins
- No trypsinization - preserve cell surface proteins
- No physical force - get high cell viability
- Releases adherent cells by reduction of temperature of the cell culture
- For culture passaging, single-cell analyses and cell transplantation research
- Enables harvesting of cell sheets and creations of 3D tissue models held together by normal cell junctions and extracellular matrix
- Minimal hands-on time
- For research and single-use only
- It is quick, clean and simple-just reduce the temperature
Note: Suitable for the harvesting of cells with intact surface proteins, for cell culture passage, single-cell analyses and cell transplantation research
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service