Skip to Content
Merck
All Photos(2)

Key Documents

Z687545

3D Biotek 3D Insert PS scaffold

for 6 well plates, 3 inserts/6 well plate

Synonym(s):

3D, 3D Cell Culture

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
$149.00

$149.00


Estimated to ship on12 April 2025



Select a Size

Change View
3 EA
$149.00

About This Item

UNSPSC Code:
41122107
NACRES:
NC.41

$149.00


Estimated to ship on12 April 2025


material

polystyrene

description

polystyrene (PS)

sterility

sterile

quality

3 inserts/6 well plate

manufacturer/tradename

3D Biotek PS152006-3

fiber diam.

~150 μm

size

6 well

color

clear

pore size

~200 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

3D Biotek offers cutting edge 3D cell culture products and technology at affordable prices. Now it is the right time to take advantage of our promotional offer and try our products. You will be amazed by how easy and convenient to use our 3D Insert cell culture scaffolds. Our 3D Insert cell culture scaffolds are:

  • Terminally sterilized by gamma irradiation and ready for use
  • Compatible with most of your current 2D assays
  • Easy for imaging. Cell growth can be monitored by light microscope, no need to purchase any additional sophisticated equipment.

Legal Information

3D Insert is a trademark of 3D Biotek, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos E Caicedo-Carvajal et al.
Journal of tissue engineering, 2011, 362326-362326 (2011-11-11)
Isolation and amplification of primary lymphoma cells in vitro setting is technically and biologically challenging task. To optimize culture environment and mimic in vivo conditions, lymphoma cell lines were used as a test case and were grown in 3-dimension (3D)

Articles

3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service