Recommended Products
manufacturer/tradename
BRAND 44850
volume
2 L
20000 mL
application(s)
detection
Looking for similar products? Visit Product Comparison Guide
General description
Ingredients:
100 g Pursept-AF contain: 12.5 g Dicedyl-dimethyl-ammoniumchloride, 10.4 g Alkylpropylenediamine-bis-guanidiniumdiacetate, 1.5 g Bis(aminopropyl)-laurylamine, auxiliary agents
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service