Skip to Content
Merck
All Photos(1)

Documents

Z4402

Sigma-Aldrich

Zinterol hydrochloride

≥98% (HPLC)

Synonym(s):

Zinterol hydrochloride, N-[5-[2-[(1,1-dimethyl-2-phenylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]-Methanesulfonamide Hydrochloride; MJ 9184

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26N2O4S·HCl
CAS Number:
Molecular Weight:
414.95
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

solubility

DMSO: ≥15 mg/mL

storage temp.

room temp

SMILES string

CC(C)(Cc1ccccc1)NCC(O)c2ccc(O)c(NS(C)(=O)=O)c2

InChI

1S/C19H26N2O4S/c1-19(2,12-14-7-5-4-6-8-14)20-13-18(23)15-9-10-17(22)16(11-15)21-26(3,24)25/h4-11,18,20-23H,12-13H2,1-3H3

InChI key

XJBCFFLVLOPYBV-UHFFFAOYSA-N

Biochem/physiol Actions

Zinterol Hydrochloride is a β2 adrenergic receptor selective agonist.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H E Lara et al.
Endocrine, 15(2), 187-192 (2001-11-27)
We investigated the possibility that norepinephrine from the human ovary is released after nerve stimulation and that this neurotransmitter is coupled to a steroidogenic response. Biologically significant levels of both norepinephrine and dopamine were found in human ovarian biopsies. [3H]norepinephrine
S Magne et al.
The Journal of biological chemistry, 276(43), 39539-39548 (2001-08-17)
We have recently reported that arachidonic acid mediates beta(2)-adrenergic receptor (AR) stimulation of [Ca(2+)](i) cycling and cell contraction in embryonic chick ventricular cardiomyocytes (Pavoine, C., Magne, S., Sauvadet, A., and Pecker, F. (1999) J. Biol. Chem. 274, 628-637). In the
Dana S Hutchinson et al.
Naunyn-Schmiedeberg's archives of pharmacology, 373(2), 158-168 (2006-04-08)
The present study investigates the action of zinterol at beta(3)-adrenoceptors. We used mouse primary brown adipocytes and Chinese hamster ovary (CHO-K1) cells expressing the mouse or human beta(3)-adrenoceptor. Zinterol was a full agonist at increasing cyclic AMP levels in primary
Ismayil Ahmet et al.
Circulation, 110(9), 1083-1090 (2004-08-18)
Studies in isolated cardiac myocytes have demonstrated that signaling via specific beta1-adrenergic receptor subtypes (beta1ARs) promotes but that signaling via beta2ARs protects from cell death. We hypothesized that prolonged beta(2)AR stimulation or beta1AR blockade would each protect myocytes from death
Susan F Steinberg et al.
American journal of physiology. Heart and circulatory physiology, 282(4), H1181-H1188 (2002-03-15)
We studied the role of beta-adrenergic receptor subtype signaling to cAMP and calcium in the genesis of catecholamine-dependent arrhythmias in German shepherd dogs that develop lethal arrhythmias at ~5 mo of age. There were three major findings in this study:

Articles

β-Adrenoceptors are activated either by norepinephrine or epinephrine. Learn more about the role of β-adrenoceptors in protein expression, cell signaling, and metabolism research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service