These cellophane sheets are approximately 1.6 mm in thickness with a pore size of approximately 4 nm which allows for water vapor.
Z377597
Gel drying frames
Extra cellophane sheets, size 14 cm × 14 cm
Synonym(s):
crack prevention gel drying rack, gel drying frame
Select a Size
$43.70
In StockDetails
Select a Size
About This Item
$43.70
In StockDetails
Recommended Products
packaging
pk of 50 sheets
manufacturer/tradename
Diversified Biotech AB-110762
size
14 cm × 14 cm
Related Categories
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
What is the dalton size of the cellophane sheet which are provided in this package and can they be used in permeability testing
1 answer-
Helpful?
-